Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037104.mm.1 |
TC0200004443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037104.mm.1 |
NM_027175.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037104.mm.1 |
NM_027175.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037106.mm.1 |
TC0200004443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037106.mm.1 |
NM_027175.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037106.mm.1 |
NM_027175.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037103.mm.1 |
TC0200004443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037103.mm.1 |
NM_027175.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037103.mm.1 |
NM_027175.3 |
Refseq |