Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333960_g1
          See other GPCPD1 GE Assays ›
          Gene Symbol:
          Gpcpd1
          Gene Name:
          glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae)
          Gene Aliases:
          2310004G06Rik, 2310032D16Rik, AU015213, Gde5, Prei4, mKIAA1434
          Chromosome Location:
          Chr.2: 132529082 - 132578282 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333960_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpcpd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042672.1 NP_001036137.1 13 - 14 1440 133
          NM_001291050.1 NP_001277979.1 12 - 13 1313 133
          NM_001291051.1 NP_001277980.1 12 - 13 1343 133
          NM_001291052.1 NP_001277981.1 12 - 13 1361 133
          NM_027096.2 NP_081372.1 11 - 12 1237 133
          NM_028802.3 NP_083078.3 13 - 14 1437 133
          GenBank mRNA AK009563.1 - 6 - 7 656 133
          AK030645.1 - 13 - 14 1391 133
          AK031658.1 - 12 - 13 1316 133
          AK049491.1 - 5 - 6 526 133
          AK050318.1 - 13 - 14 1352 133
          AK051728.1 - 5 - 6 500 133
          AK141374.1 - 13 - 14 1327 133
          AK163419.1 - 12 - 13 1284 133
          AK165952.1 - 13 - 14 1402 133
          AK166293.1 - 4 - 5 371 133
          AK166454.1 - 12 - 13 1326 133
          BC006887.1 - 12 - 13 1285 133
          BC033408.1 - 11 - 12 1167 5357683 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004676.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpcpd1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          skeletal muscle tissue development
          phosphoric diester hydrolase activity
          hydrolase activity
          carbohydrate binding
          glycerophosphocholine phosphodiesterase activity
          starch binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200040166.mm.1 TC0200004676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040166.mm.1 NM_001042672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040166.mm.1 NM_001042672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040166.mm.1 NM_028802.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0