Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01334168_m1
          See other NXT1 GE Assays ›
          Gene Symbol:
          Nxt1
          Gene Name:
          NTF2-related export protein 1
          Gene Aliases:
          1110001N02Rik
          Chromosome Location:
          Chr.2: 148672615 - 148676026 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01334168_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110159.1 NP_001103629.1 2 - 3 337 69
          GenBank mRNA BY091455.1 - 2 - 3 321 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001110159.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          RNA export from nucleus
          protein export from nucleus
          transport
          protein transport
          mRNA transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731512 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064811 BC023672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064811 NM_001110159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064811 NM_019761.6 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064811 AW908959.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77719 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143449 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064811 BC023672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064811 NM_001110159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064811 NM_019761.6 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064811 AW908959.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184398 CJ186513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184398 BC023672.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184398 NM_001110159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184398 NM_019761.6 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184398 AF156958.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413419 BC023672.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413419 NM_001110159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413419 NM_019761.6 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413419 AF156958.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413419 AK168314.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21731508 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04734150 - -
          Whole Mouse Genome G4122A A_51_P413419 BC023672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413419 NM_001110159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413419 NM_019761.6 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413419 AF156958.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413419 AK168314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184398 CJ186513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184398 BC023672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184398 NM_001110159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184398 NM_019761.6 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184398 AF156958.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124826 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04734150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359875 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359864 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01359878 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21731511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21731570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02242390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01837000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01359872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01359865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01605960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01474170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01359868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01605961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01782367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01782354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01782361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21731508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01605962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01782356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01359864 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10476892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10476890 - -
          Affymetrix HT MG-430 PM Array Plate 1422488_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10476889 - -
          Mouse Genome 430 2.0 Array 1422488_at - -
          Mouse Genome 430 2.0 Array 1422488_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56488_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56488_at - -
          Mouse Expression Set 430 1422488_at - -
          Mouse Expression Set 430 1422488_PM_at - -
          Murine Genome U74v2 Set 160429_at - -
          Murine Genome U74v2 Set 160429_PM_at - -
          Murine Genome U74v2 Set 167719_PM_f_at - -
          Murine Genome U74v2 Set 167719_f_at - -
          Mouse Genome 430A 2.0 Array 1422488_PM_at - -
          Mouse Genome 430A 2.0 Array 1422488_at - -
          Affymetrix HT Mouse Genome 430A Array 1422488_at - -
          Affymetrix HT Mouse Genome 430A Array 1422488_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6400037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234152 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234152 - -
          mouse-6 v1.1 (Illumina) scl20160.1.79_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20160.1.79_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2975041 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2975041 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline