Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335332_m1
          See other 9030622O22RIK GE Assays ›
          Gene Symbol:
          9030622O22Rik
          Gene Name:
          RIKEN cDNA 9030622O22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 147944459 - 148040560 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335332_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9030622O22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK018565.1 - 6 - 7 959 109

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA A530006G24 gene A530006G24Rik Gm1003 -227.2kb Anti-sense Chr. 2 - 147710693 - 147717632 on NCBI Build GRCm38
          Downstream forkhead box A2 Foxa2 HNF3-beta,
          HNF3beta,
          Hnf-3b,
          Hnf3b,
          Tcf-3b,
          Tcf3b
          95.7kb Sense Chr. 2 - 148042877 - 148046969 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097964 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097962 AK018565.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 236888 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44843 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097962 AK018565.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01365995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02018674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02018661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02123779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01749186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02018656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02123782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02018671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01665121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01960256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02018685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02123760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01996856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02123778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21727359 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431566_PM_at - -
          Mouse Genome 430 2.0 Array 1431566_at - -
          Mouse Genome 430 2.0 Array 1431566_PM_at - -
          Mouse Expression Set 430 1431566_at - -
          Mouse Expression Set 430 1431566_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431566_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431566_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0