Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335369_m1
          See other 1700016G22RIK GE Assays ›
          Gene Symbol:
          1700016G22Rik
          Gene Name:
          RIKEN cDNA 1700016G22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 5855509 - 5858092 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335369_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700016G22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045891.1 2 - 3 149 131
          GenBank mRNA AK006031.1 - 2 - 3 151 131

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream aldo-keto reductase family 1,
          member E1
          Akr1e1 1810061I10Rik,
          Akr1e2
          -1.2mb Sense Chr. 13 - 4592490 - 4609164 on NCBI Build GRCm38
          Downstream Kruppel-like factor 6 Klf6 AI448727,
          BCD1,
          C86813,
          CPBP,
          Copeb,
          FM2,
          FM6,
          Ierepo1,
          Ierepo3,
          R75280,
          Zf9
          4.6kb Anti-sense Chr. 13 - 5861489 - 5870393 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001389.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2044877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922508 - -
          Whole Mouse Genome G4122A A_52_P1098820 AK006031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1098820 AV045830.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10969595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10959736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10501423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28798784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300008705.mm.1 TC1300001389.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008705.mm.1 NR_045891.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008705.mm.1 NR_045891.1 Refseq
          Murine Genome U74v2 Set 169325_PM_i_at - -
          Murine Genome U74v2 Set 169325_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358895 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102060102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0