Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1430939_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430939_at |
- |
- |
Mouse Genome 430 2.0 Array |
1430939_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1300004830.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004830.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004830.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004838.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004838.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004838.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004839.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004839.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004839.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004832.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004832.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004832.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004833.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004833.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004833.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004829.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004829.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004829.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004831.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004831.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004831.mm.1 |
NR_045977.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004834.mm.1 |
TC1300000727.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300004834.mm.1 |
NR_045977.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300004834.mm.1 |
NR_045977.1 |
Refseq |
Mouse Expression Set 430 |
1430939_PM_at |
- |
- |
Mouse Expression Set 430 |
1430939_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430939_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1430939_PM_at |
- |
- |