Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335859_m1
          See other GM9962 GE Assays ›
          Gene Symbol:
          Gm9962
          Gene Name:
          predicted gene 9962
          Gene Aliases:
          ENSMUSG00000055048
          Chromosome Location:
          Chr.7: 57387272 - 57409941 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335859_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm9962
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033504.1 2 - 3 237 67
          GenBank mRNA AK049950.1 - 2 - 3 238 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB653224.1 - 2 - 3 238 Chr. 7 - 57387272 - 57409168 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream gamma-aminobutyric acid (GABA) A receptor,
          subunit gamma 3
          Gabrg3 B230362M20Rik,
          Gabrg-3
          -17.0kb Anti-sense Chr. 7 - 56724242 - 57386871 on NCBI Build GRCm38
          Downstream microRNA 6389 Mir6389 Gm27946,
          mmu-mir-6389
          171.1kb Sense Chr. 7 - 57581061 - 57581162 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000939.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038559 AK049950.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038559 NR_033504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038559 BC062112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038559 AK038476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038559 AK083185.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158604 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131567 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038559 AK049950.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038559 NR_033504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038559 BC062112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038559 AK038476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2038559 AK083185.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06207776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158604 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119501 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158604 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25150598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25150599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06625293 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10553769 - -
          Affymetrix Mouse Clariom S Array PSR0700008654.mm.1 TC0700000939.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008654.mm.1 NR_033504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008654.mm.1 NR_033504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008655.mm.1 TC0700000939.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700008655.mm.1 NR_033504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700008655.mm.1 NR_033504.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 789067 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103360390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240151 - -
          Illumina Sentrix mouse-6.v1 bead array 103360390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103360390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0