Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200023940.mm.1 |
TC0200002883.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023940.mm.1 |
NM_021305.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023940.mm.1 |
NM_021305.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023942.mm.1 |
TC0200002883.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023942.mm.1 |
NM_021305.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023942.mm.1 |
NM_021305.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023936.mm.1 |
TC0200002883.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023936.mm.1 |
NM_021305.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023936.mm.1 |
NM_021305.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023939.mm.1 |
TC0200002883.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023939.mm.1 |
NM_021305.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023939.mm.1 |
NM_021305.3 |
Refseq |