Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016719.1 | NP_057928.1 | 1 - 2 | 299 | 73 | |
GenBank mRNA | AF155647.1 | - | 1 - 2 | 299 | 73 | |
AK010849.1 | - | 1 - 2 | 316 | 73 | ||
AK010903.1 | - | 1 - 2 | 316 | 73 | ||
AK147041.1 | - | 1 - 2 | 308 | 73 | ||
BC138581.1 | - | 1 - 2 | 205 | 8861601 | 73 | |
BC145913.1 | - | 1 - 2 | 205 | 40131216 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |