Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01337978_g1
          See other RMDN3 GE Assays ›
          Gene Symbol:
          Rmdn3
          Gene Name:
          regulator of microtubule dynamics 3
          Gene Aliases:
          1200015F23Rik, AI131757, Fam82a2, Ptpip51, RMD-3, Rmd3
          Chromosome Location:
          Chr.2: 119136998 - 119157034 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01337978_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rmdn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033136.3 NP_001028308.1 12 - 13 1502 100
          GenBank mRNA AK004786.1 - 12 - 13 1428 100
          AK146299.1 - 12 - 13 1421 100
          AK151090.1 - 12 - 13 1423 100
          AK153322.1 - 12 - 13 1423 100
          AK153369.1 - 12 - 13 1422 100
          AK171770.1 - 12 - 13 1503 100
          BC055754.1 - 12 - 13 1396 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004423.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          apoptotic process
          cell differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 BC055754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK151090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK153322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK153369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 NM_001033136.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 BC055754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK151090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK153322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 AK153369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233027 NM_001033136.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 AK010581.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 AK145843.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 AK168384.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 AK178869.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 AK183750.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 BC086644.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 BC131945.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 BC131973.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 D82983.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 NM_008820.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735371 U51014.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198838 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 55591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 17489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731262 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22567 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144128 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110136 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 BC055754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK151090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK153322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK153369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 NM_001033136.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 BC055754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK151090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK153322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 AK153369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233027 NM_001033136.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21568172 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144128 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110136 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04664392 - -
          Whole Mouse Genome G4122A A_52_P333749 BC055754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333749 AK151090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333749 AK153322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333749 AK153369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333749 NM_001033136.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333806 BC055754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333806 AK151090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333806 AK153322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333806 AK153369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P333806 NM_001033136.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 BC055754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK151090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK153322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK153369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 NM_001033136.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 BC055754.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK151090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK153322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 AK153369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233027 NM_001033136.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01820843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21567981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21568067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02021291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02021296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02021292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21568052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10486173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10486181 - -
          Affymetrix HT MG-430 PM Array Plate 1452704_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428196_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10486172 - -
          Mouse Genome 430 2.0 Array 1428196_a_at - -
          Mouse Genome 430 2.0 Array 1452704_at - -
          Mouse Genome 430 2.0 Array 1452704_PM_at - -
          Mouse Genome 430 2.0 Array 1458722_at - -
          Mouse Genome 430 2.0 Array 1428196_PM_a_at - -
          Mouse Genome 430 2.0 Array 1458722_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67809_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67809_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200036928.mm.1 TC0200004423.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036928.mm.1 NM_001033136.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036928.mm.1 NM_001033136.3 Refseq
          Mouse Expression Set 430 1452704_at - -
          Mouse Expression Set 430 1428196_PM_a_at - -
          Mouse Expression Set 430 1452704_PM_at - -
          Mouse Expression Set 430 1428196_a_at - -
          Murine Genome U74v2 Set 95078_PM_at - -
          Murine Genome U74v2 Set 95078_at - -
          Murine Genome U74v2 Set 169610_PM_r_at - -
          Murine Genome U74v2 Set 169610_r_at - -
          Mouse Genome 430A 2.0 Array 1452704_at - -
          Mouse Genome 430A 2.0 Array 1428196_a_at - -
          Mouse Genome 430A 2.0 Array 1452704_PM_at - -
          Mouse Genome 430A 2.0 Array 1428196_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452704_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428196_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428196_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452704_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2753515 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2753515 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3071764 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3150536 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3150536 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3071764 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0