Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338020_m1
          See other DLL4 GE Assays ›
          Gene Symbol:
          Dll4
          Gene Name:
          delta-like 4 (Drosophila)
          Gene Aliases:
          Delta4
          Chromosome Location:
          Chr.2: 119325784 - 119335666 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338020_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dll4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019454.3 NP_062327.2 9 - 10 2302 68
          GenBank mRNA AB043893.1 - 9 - 10 1945 68
          AF253469.1 - 9 - 10 2257 68
          AK004739.1 - 9 - 10 2270 68
          AK031699.1 - 9 - 10 2272 68
          AK052322.1 - 9 - 10 2304 68
          AK165517.1 - 9 - 10 2270 68
          AK171717.1 - 9 - 10 2269 68
          BC042497.1 - 9 - 10 2257 4017786 68
          BC049130.1 - 9 - 10 2262 6825525 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001769.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dll4

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          branching involved in blood vessel morphogenesis
          blood vessel remodeling
          cardiac ventricle morphogenesis
          cardiac atrium morphogenesis
          ventricular trabecula myocardium morphogenesis
          pericardium morphogenesis
          cell communication
          Notch signaling pathway
          multicellular organism development
          nervous system development
          visual perception
          negative regulation of cell population proliferation
          negative regulation of endothelial cell migration
          positive regulation of gene expression
          negative regulation of gene expression
          cell differentiation
          dorsal aorta morphogenesis
          cellular response to vascular endothelial growth factor stimulus
          negative regulation of blood vessel endothelial cell migration
          cellular response to fibroblast growth factor stimulus
          regulation of neurogenesis
          response to stimulus
          ventral spinal cord interneuron fate commitment
          regulation of neural retina development
          Notch signaling involved in heart development
          blood vessel lumenization
          negative regulation of cell migration involved in sprouting angiogenesis
          negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          positive regulation of neural precursor cell proliferation
          Notch binding
          calcium ion binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200013277.mm.1 TC0200001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013277.mm.1 NM_019454.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013277.mm.1 NM_019454.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline