Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338533_m1
          See other SERPINA3J GE Assays ›
          Gene Symbol:
          Serpina3j
          Gene Name:
          serine (or cysteine) peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3J
          Gene Aliases:
          AI506554, EG238395, Gm4931
          Chromosome Location:
          Chr.12: 104300722 - 104320586 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338533_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpina3j
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101472.2 NP_001094942.1 1 - 2 633 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001043.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          negative regulation of endopeptidase activity
          molecular_function
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064989 NM_001101472.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887496 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143798 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2064989 NM_001101472.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221067 NM_001101472.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28702643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09892766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09892771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10294858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10142926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10294861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10354507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10396326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10354492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10354493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10396318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10142924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28702635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10354504 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200007462.mm.1 TC1200001043.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007462.mm.1 NM_001101472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007462.mm.1 NM_001101472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007463.mm.1 TC1200001043.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007463.mm.1 NM_001101472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007463.mm.1 NM_001101472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007461.mm.1 TC1200001043.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007461.mm.1 NM_001101472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007461.mm.1 NM_001101472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007460.mm.1 TC1200001043.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200007460.mm.1 NM_001101472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200007460.mm.1 NM_001101472.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 484615 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226074 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0