Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01339632_s1
          See other SRY GE Assays ›
          Gene Symbol:
          Sry
          Gene Name:
          sex determining region of Chr Y
          Gene Aliases:
          Tdf, Tdy
          Chromosome Location:
          Chr.Y: 2662471 - 2663658 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01339632_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sry
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011564.1 NP_035694.1 1 1160 68
          GenBank mRNA BC111528.2 - 2 1097 40045188 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0Y00000240.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sry

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          sex determination
          sex differentiation
          male gonad development
          negative regulation of gene expression
          cell differentiation
          male sex determination
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          protein binding
          calmodulin binding
          DNA binding, bending
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029637 NM_011564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029637 BC111528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029637 NM_011564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029637 BC111528.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252126 NM_011564.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P252126 BC111528.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32829278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09498009 - -
          Whole Mouse Genome G4122A A_51_P252126 NM_011564.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P252126 BC111528.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134122 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126829 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09498009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15226515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15226506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15228604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15228607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15226512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15226509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15226510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32829257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32829257 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10608194 - -
          Affymetrix Mouse Gene 1.0 ST Array 10608195 - -
          Affymetrix HT MG-430 PM Array Plate 1450579_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1450578_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10608193 - -
          Mouse Genome 430 2.0 Array 1450578_PM_at - -
          Mouse Genome 430 2.0 Array 1450579_PM_x_at - -
          Mouse Genome 430 2.0 Array 1450579_x_at - -
          Mouse Genome 430 2.0 Array 1450578_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21674_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21674_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0Y00001471.mm.1 TC0Y00000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001471.mm.1 NM_011564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001471.mm.1 NM_011564.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001470.mm.1 TC0Y00000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0Y00001470.mm.1 NM_011564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0Y00001470.mm.1 NM_011564.1 Refseq
          Mouse Expression Set 430 1450578_at - -
          Mouse Expression Set 430 1450579_PM_x_at - -
          Mouse Expression Set 430 1450579_x_at - -
          Mouse Expression Set 430 1450578_PM_at - -
          Murine Genome U74v2 Set 99556_PM_s_at - -
          Murine Genome U74v2 Set 99556_s_at - -
          Mouse Genome 430A 2.0 Array 1450579_x_at - -
          Mouse Genome 430A 2.0 Array 1450579_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1450578_PM_at - -
          Mouse Genome 430A 2.0 Array 1450578_at - -
          Affymetrix HT Mouse Genome 430A Array 1450578_at - -
          Affymetrix HT Mouse Genome 430A Array 1450579_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450579_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1450578_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 863912 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101340044 - -
          Illumina mouse-6 v1.0 expression beadchip 101340044 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2542816 - -
          mouse-6 v1.1 (Illumina) GI_6755760-S - -
          Illumina Sentrix mouse-6.v1 bead array 101340044 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101340044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255639 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255639 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0