Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01340846_m1
          See other HNRNPH2 GE Assays ›
          Gene Symbol:
          Hnrnph2
          Gene Name:
          heterogeneous nuclear ribonucleoprotein H2
          Gene Aliases:
          DXHXS1271E, Ftp-3, Ftp3, H', HNRNP, Hnrph2
          Chromosome Location:
          Chr.X: 134601184 - 134607060 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01340846_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnph2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019868.3 NP_063921.1 3 - 4 170 64
          GenBank mRNA AK077716.1 - 3 - 4 171 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001283.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK077716.1, NM_019868.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hnrnph2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672784 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672783 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133895 BC005461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133895 AK168116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133895 AK169104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133895 NM_019868.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133895 AK077716.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325996 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327667 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159624 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133895 BC005461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133895 AK168116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133895 AK169104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133895 NM_019868.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133895 AK077716.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32672789 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159624 - -
          Whole Mouse Genome G4122A A_51_P333885 BC005461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333885 AK168116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333885 AK169104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333885 NM_019868.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333885 AK077716.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126073 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14846954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672782 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672783 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672784 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32672788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32672788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15209891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15213568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14846944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14990827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14961170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15208867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14844097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14835287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32672808 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10601763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10601761 - -
          Affymetrix HT MG-430 PM Array Plate 1415963_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10490773 - -
          Affymetrix Mouse Gene 1.1 ST Array 10601760 - -
          Mouse Genome 430 2.0 Array 1415963_PM_at - -
          Mouse Genome 430 2.0 Array 1415963_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008018.mm.1 TC0X00001283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008018.mm.1 NM_019868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008020.mm.1 TC0X00001283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008020.mm.1 NM_019868.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008019.mm.1 TC0X00001283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008019.mm.1 NM_019868.4 Refseq
          Mouse Expression Set 430 1415963_at - -
          Mouse Expression Set 430 1415963_PM_at - -
          Murine Genome U74v2 Set 94832_PM_at - -
          Murine Genome U74v2 Set 94832_at - -
          Mouse Genome 430A 2.0 Array 1415963_PM_at - -
          Mouse Genome 430A 2.0 Array 1415963_at - -
          Affymetrix HT Mouse Genome 430A Array 1415963_at - -
          Affymetrix HT Mouse Genome 430A Array 1415963_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2630040 - -
          Illumina mouse-6 v1.0 expression beadchip 2630040 - -
          Illumina mouse-6 v1.0 expression beadchip 5720671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680808 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596458 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680808 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596458 - -
          mouse-6 v1.1 (Illumina) scl0056258.1_110-S - -
          mouse-6 v1.1 (Illumina) scl056258.4_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630040 - -
          Illumina Sentrix mouse-6.v1 bead array 5720671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056258.4_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056258.1_110-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630040 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2680808 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680808 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0056258.1_110-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0