Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012056.2 | NP_036186.2 | 2 - 3 | 516 | 118 | |
GenBank mRNA | AF090334.1 | - | 2 - 3 | 442 | 118 | |
AF279263.1 | - | 2 - 3 | 494 | 118 | ||
AK198658.1 | - | 2 - 3 | 186 | 118 | ||
AK204047.1 | - | 2 - 3 | 186 | 118 | ||
AK206852.1 | - | 1 - 2 | 67 | 118 | ||
AK208794.1 | - | 2 - 3 | 183 | 118 | ||
AK209926.1 | - | 1 - 2 | 136 | 118 | ||
AK214409.1 | - | 2 - 3 | 186 | 118 | ||
AK216520.1 | - | 2 - 3 | 186 | 118 | ||
BC043129.1 | - | 2 - 3 | 516 | 6810029 | 118 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |