Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900013913.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013913.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013913.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013909.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013909.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013909.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013912.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013912.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013912.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013911.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013911.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013911.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013908.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013908.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013908.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013910.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013910.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013910.mm.1 |
NM_177911.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013905.mm.1 |
TC0900001655.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900013905.mm.1 |
NM_177911.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900013905.mm.1 |
NM_177911.4 |
Refseq |