Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01341557_m1
          See other 4930412L05RIK GE Assays ›
          Gene Symbol:
          4930412L05Rik
          Gene Name:
          RIKEN cDNA 4930412L05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 53201777 - 53220013 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01341557_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930412L05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015122.1 - 7 - 8 1008 129

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATP-binding cassette,
          sub-family A (ABC1),
          member 1
          Abca1 ABC-1,
          Abc1
          -45.1kb Sense Chr. 4 - 53030789 - 53159988 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930522O17 gene 4930522O17Rik 39.0kb Anti-sense Chr. 4 - 53246196 - 53251120 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2299186 AK141187.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138593 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132620 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123464 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04062130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03894195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03316128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03602268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03783763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03538895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03316130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03884783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03894194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03856777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04062137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03316125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04062120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03263951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04100621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03316117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03869731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03403295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03856766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03316122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04100645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03856784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03485269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04062122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22922006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04100619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22921845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04100649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04026992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432193_PM_at - -
          Mouse Genome 430 2.0 Array 1432193_PM_at - -
          Mouse Genome 430 2.0 Array 1432193_at - -
          Mouse Expression Set 430 1432193_PM_at - -
          Mouse Expression Set 430 1432193_at - -
          Affymetrix HT Mouse Genome 430B Array 1432193_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432193_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103360446 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0