Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01342088_g1
          See other ATF7IP2 GE Assays ›
          Gene Symbol:
          Atf7ip2
          Gene Name:
          activating transcription factor 7 interacting protein 2
          Gene Aliases:
          4930558K11Rik, BC018510, Get-1, PSM2
          Chromosome Location:
          Chr.16: 10192640 - 10249669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01342088_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atf7ip2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029253.1 NP_083529.1 8 - 9 921 141
          NM_153123.2 NP_694763.2 8 - 9 921 141
          GenBank mRNA AK016180.1 - 8 - 9 923 141
          AK077229.1 - 8 - 9 881 141
          AK145634.1 - 8 - 9 880 141
          AY903215.1 - 8 - 9 905 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000110.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atf7ip2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 BC018510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 AK145634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 NM_153123.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 BC018510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 AK145634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P378511 NM_153123.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005893 AK145634.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005893 AY903215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005893 AK077229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005893 NM_153123.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005893 AK016180.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633816 BC076625.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633816 BC085176.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633816 DQ192038.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633816 NM_001007580.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89448 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108948 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 895586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387722 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 95413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 147097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144653 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177887 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168884 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180512 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005893 AK145634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005893 AY903215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005893 AK077229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005893 NM_153123.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005893 AK016180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 BC018510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 AK145634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 NM_153123.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 BC018510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 AK145634.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P378511 NM_153123.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177887 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168884 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08505940 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08505962 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08505917 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08505909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180512 - -
          Whole Mouse Genome G4122A A_52_P247233 AK049401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P620766 BY715784.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P620766 AK139862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P620766 AK145634.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P620766 AK077229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P620766 NM_153123.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196148 AY903215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196148 AK077229.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08505940 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180512 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168884 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12538590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12651199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12656454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12920797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12920796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12597686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30541244 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433489 - -
          Affymetrix HT MG-430 PM Array Plate 1442796_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430364_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427603_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433484 - -
          Affymetrix Mouse Gene 1.1 ST Array 10433492 - -
          Mouse Genome 430 2.0 Array 1427603_at - -
          Mouse Genome 430 2.0 Array 1427603_PM_at - -
          Mouse Genome 430 2.0 Array 1442796_at - -
          Mouse Genome 430 2.0 Array 1430364_PM_at - -
          Mouse Genome 430 2.0 Array 1442796_PM_at - -
          Mouse Genome 430 2.0 Array 1430364_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75329_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75329_at - -
          Affymetrix Mouse Clariom S Array PSR1600000763.mm.1 TC1600000110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000763.mm.1 NM_029253.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000763.mm.1 NM_029253.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000762.mm.1 TC1600000110.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000762.mm.1 NM_029253.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000762.mm.1 NM_029253.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000762.mm.1 NM_153123.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000762.mm.1 NM_153123.2 Refseq
          Mouse Expression Set 430 1442796_at - -
          Mouse Expression Set 430 1442796_PM_at - -
          Mouse Expression Set 430 1430364_PM_at - -
          Mouse Expression Set 430 1430364_at - -
          Murine Genome U74v2 Set 168286_PM_i_at - -
          Murine Genome U74v2 Set 168286_i_at - -
          Affymetrix HT Mouse Genome 430A Array 1427603_at - -
          Affymetrix HT Mouse Genome 430A Array 1427603_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442796_at - -
          Affymetrix HT Mouse Genome 430B Array 1430364_at - -
          Affymetrix HT Mouse Genome 430B Array 1442796_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430364_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290215 - -
          Illumina mouse-6 v1.0 expression beadchip 107050390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687679 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620418 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687679 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229033 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620418 - -
          mouse-6 v1.1 (Illumina) scl0223963.1_197-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290215 - -
          Illumina Sentrix mouse-6.v1 bead array 107050390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0223963.1_197-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107050390 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620418 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620418 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229033 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline