Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600012939.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012939.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012939.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012939.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012939.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012935.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012935.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012935.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012935.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012935.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012938.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012938.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012938.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012938.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012938.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012931.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012931.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012931.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012931.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012931.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012932.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012932.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012932.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012932.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012932.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012934.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012934.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012934.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012934.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012934.mm.1 |
NM_021301.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012937.mm.1 |
TC1600001582.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600012937.mm.1 |
NM_001145899.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012937.mm.1 |
NM_001145899.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012937.mm.1 |
NM_021301.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600012937.mm.1 |
NM_021301.3 |
Refseq |