Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01343854_m1
          See other 1700026J12RIK GE Assays ›
          Gene Symbol:
          1700026J12Rik
          Gene Name:
          RIKEN cDNA 1700026J12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 49210640 - 49255798 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01343854_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700026J12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006389.1 - 7 - 8 893 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream myosin,
          heavy chain 15
          Myh15 EG667772 -11.5kb Anti-sense Chr. 16 - 49057486 - 49199104 on NCBI Build GRCm38
          Downstream intraflagellar transport 57 Ift57 4833420A15Rik,
          Esrrbl1,
          Hippi,
          MHS4R2
          482.4kb Anti-sense Chr. 16 - 49699294 - 49765126 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983354 AK006389.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42356 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101446 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144179 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P107959 AK006389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983354 AK006389.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08593304 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101446 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08593278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144179 - -
          Whole Mouse Genome G4122A A_52_P72572 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P107959 AK006389.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144179 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12878945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13008913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12539182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12626460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12721643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30744883 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453793_PM_at - -
          Mouse Genome 430 2.0 Array 1453793_at - -
          Mouse Genome 430 2.0 Array 1453793_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73255_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73255_at - -
          Mouse Expression Set 430 1453793_PM_at - -
          Mouse Expression Set 430 1453793_at - -
          Murine Genome U74v2 Set 133351_PM_at - -
          Murine Genome U74v2 Set 133351_at - -
          Affymetrix HT Mouse Genome 430B Array 1453793_at - -
          Affymetrix HT Mouse Genome 430B Array 1453793_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230603 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0