Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344347_g1
          See other NTAN1 GE Assays ›
          Gene Symbol:
          Ntan1
          Gene Name:
          N-terminal Asn amidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 13818834 - 13835450 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344347_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ntan1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010946.3 NP_035076.1 9 - 10 831 59
          GenBank mRNA AK041112.1 - 9 - 10 788 59
          BC030172.1 - 9 - 10 829 5037501 59
          U57692.1 - 9 - 10 785 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000167.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ntan1

          Gene Ontology Categories:

          Function(s) Process(es)

          memory
          adult locomotory behavior
          histone deacetylase activity
          protein-N-terminal asparagine amidohydrolase activity
          UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
          hydrolase activity
          iprodione amidohydrolase activity
          (3,5-dichlorophenylurea)acetate amidohydrolase activity
          4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
          didemethylisoproturon amidohydrolase activity
          N-isopropylacetanilide amidohydrolase activity
          N-cyclohexylformamide amidohydrolase activity
          isonicotinic acid hydrazide hydrolase activity
          cis-aconitamide amidase activity
          gamma-N-formylaminovinylacetate hydrolase activity
          O-succinylbenzoate synthase activity
          indoleacetamide hydrolase activity
          N-acetylcitrulline deacetylase activity
          N-acetylgalactosamine-6-phosphate deacetylase activity
          diacetylchitobiose deacetylase activity
          chitooligosaccharide deacetylase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559117 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559118 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559120 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559116 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559114 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559119 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158319 AK163696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158319 AK054329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158319 AK004611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158319 NM_001039533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158319 BC030172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117319 BC030172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117319 NM_010946.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117319 AA161752.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337243 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134991 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126439 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112075 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117319 BC030172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117319 NM_010946.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117319 AA161752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158319 AK163696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158319 AK054329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158319 AK004611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158319 NM_001039533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158319 BC030172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P566579 AK214245.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P566579 BC030172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P566579 AK041112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P566579 AK207258.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P566579 U57692.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30559105 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03135825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08513819 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112075 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126439 - -
          Whole Mouse Genome G4122A A_52_P566579 AK214245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566579 BC030172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566579 AK041112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566579 AK207258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P566579 U57692.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133613 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126439 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12780195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12780194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12726044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12726045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559116 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559120 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559118 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559117 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30559114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30559140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12473496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12726045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12858480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12780195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12927747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12915619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12940251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12706604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30559100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12764452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12597141 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433699 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433695 - -
          Affymetrix HT MG-430 PM Array Plate 1423538_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433691 - -
          Mouse Genome 430 2.0 Array 1423538_at - -
          Mouse Genome 430 2.0 Array 1447917_PM_x_at - -
          Mouse Genome 430 2.0 Array 1447917_x_at - -
          Mouse Genome 430 2.0 Array 1428200_PM_a_at - -
          Mouse Genome 430 2.0 Array 1423538_PM_at - -
          Mouse Genome 430 2.0 Array 1428200_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18203_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18203_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600001204.mm.1 TC1600000167.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001204.mm.1 NM_010946.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001203.mm.1 TC1600000167.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001203.mm.1 NM_010946.4 Refseq
          Mouse Expression Set 430 1423538_at - -
          Mouse Expression Set 430 1423538_PM_at - -
          Mouse Expression Set 430 1447917_x_at - -
          Mouse Expression Set 430 1447917_PM_x_at - -
          Murine Genome U74v2 Set 162083_PM_f_at - -
          Murine Genome U74v2 Set 96666_PM_at - -
          Murine Genome U74v2 Set 162083_f_at - -
          Murine Genome U74v2 Set 96666_at - -
          Mouse Genome 430A 2.0 Array 1423538_at - -
          Mouse Genome 430A 2.0 Array 1423538_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423538_at - -
          Affymetrix HT Mouse Genome 430A Array 1423538_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447917_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447917_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3440593 - -
          Illumina mouse-6 v1.0 expression beadchip 3440593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254516 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254516 - -
          mouse-6 v1.1 (Illumina) scl49405.9.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49405.9.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2888742 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254516 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888742 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49405.9.1_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline