Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344380_g1
          See other PDXDC1 GE Assays ›
          Gene Symbol:
          Pdxdc1
          Gene Name:
          pyridoxal-dependent decarboxylase domain containing 1
          Gene Aliases:
          2210010A19Rik, AA415817, Kiaa0251-hp
          Chromosome Location:
          Chr.16: 13833161 - 13903145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344380_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdxdc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039533.2 NP_001034622.1 18 - 19 1903 147
          NM_001291017.1 NP_001277946.1 18 - 19 1903 147
          NM_053181.3 NP_444411.2 18 - 19 1903 147
          GenBank mRNA AK004611.1 - 18 - 19 1780 147
          AK030613.1 - 18 - 19 1734 147
          AK033107.1 - 18 - 19 1734 147
          AK054329.1 - 18 - 19 1886 147
          AK075829.1 - 11 - 12 1026 147
          AK163696.1 - 18 - 19 1774 147
          AK165587.1 - 18 - 19 1738 147
          AK178472.1 - 3 - 4 147
          BC005541.1 - 18 - 19 1721 3590316 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001236.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdxdc1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          carboxylic acid metabolic process
          catalytic activity
          phosphoenolpyruvate carboxykinase activity
          gamma-glutamyl carboxylase activity
          3-octaprenyl-4-hydroxybenzoate carboxy-lyase activity
          lyase activity
          carboxy-lyase activity
          2-hydroxy-3-carboxy-6-oxo-7-methylocta-2,4-dienoate decarboxylase activity
          bis(4-chlorophenyl)acetate decarboxylase activity
          3,5-dibromo-4-hydroxybenzoate decarboxylase activity
          2-hydroxyisobutyrate decarboxylase activity
          2-hydroxy-2-methyl-1,3-dicarbonate decarboxylase activity
          pyridoxal phosphate binding
          2-hydroxyisophthalate decarboxylase activity
          dimethylmalonate decarboxylase activity
          2,4,4-trimethyl-3-oxopentanoate decarboxylase activity
          4,4-dimethyl-3-oxopentanoate decarboxylase activity
          2,3,6-trihydroxyisonicotinate decarboxylase activity
          phenanthrene-4,5-dicarboxylate decarboxylase activity
          pyrrole-2-carboxylate decarboxylase activity
          terephthalate decarboxylase activity
          malonate semialdehyde decarboxylase activity
          5-amino-4-imidazole carboxylate lyase activity
          2-keto-4-methylthiobutyrate aminotransferase activity
          2-oxo-4-hydroxy-4-carboxy-5-ureidoimidazoline decarboxylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600009897.mm.1 TC1600001236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009897.mm.1 NM_001039533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009897.mm.1 NM_053181.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009896.mm.1 TC1600001236.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600009896.mm.1 NM_001039533.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600009896.mm.1 NM_053181.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline