Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344947_m1
          See other PPM1F GE Assays ›
          Gene Symbol:
          Ppm1f
          Gene Name:
          protein phosphatase 1F (PP2C domain containing)
          Gene Aliases:
          1110021B16Rik, 4933427B07Rik, CaMKPase, Popx2, mKIAA0015
          Chromosome Location:
          Chr.16: 16896469 - 16927375 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344947_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppm1f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176833.4 NP_789803.1 4 - 5 770 77
          GenBank mRNA AK016942.1 - 4 - 5 775 77
          AK154192.1 - 4 - 5 673 77
          AK164964.1 - 4 - 5 651 77
          BC042570.1 - 3 - 4 619 4241448 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000201.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppm1f

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          protein dephosphorylation
          apoptotic process
          positive regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of cell-substrate adhesion
          histone dephosphorylation
          negative regulation of peptidyl-serine phosphorylation
          peptidyl-threonine dephosphorylation
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          negative regulation of transcription, DNA-templated
          positive regulation of growth
          positive regulation of chemotaxis
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          cellular response to xenobiotic stimulus
          intrinsic apoptotic signaling pathway
          catalytic activity
          phosphoprotein phosphatase activity
          protein serine/threonine phosphatase activity
          hydrolase activity
          phosphatase activity
          calmodulin-dependent protein phosphatase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600001608.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001608.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001608.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001607.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001607.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001607.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001605.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001605.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001605.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001603.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001603.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001603.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001609.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001609.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001609.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001602.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001602.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001602.mm.1 NM_176833.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001606.mm.1 TC1600000201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001606.mm.1 NM_176833.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001606.mm.1 NM_176833.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0