Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01345074_m1
          See other LRRC74B GE Assays ›
          Gene Symbol:
          Lrrc74b
          Gene Name:
          leucine rich repeat containing 74B
          Gene Aliases:
          4930451C15Rik, mCG12354, mCP17390
          Chromosome Location:
          Chr.16: 17544465 - 17561368 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01345074_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc74b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145435.1 NP_001138907.1 5 - 6 757 85
          NM_029053.3 NP_083329.2 5 - 6 757 85
          GenBank mRNA AK015438.1 - 2 - 3 1231 85
          AK039092.1 - 5 - 6 735 85
          AK132588.1 - 5 - 6 755 85
          AK143245.1 - 5 - 6 730 85
          AK161226.1 - 5 - 6 755 85
          BC115678.1 - 5 - 6 734 40104166 85
          BC115679.1 - 5 - 6 734 40104167 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001293.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK015438.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK161226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 BC115678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 BC115679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK132588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK143245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK161226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 BC115678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 BC115679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK132588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P347764 AK143245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006183 AK039092.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117720 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167024 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161918 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK161226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 BC115678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 BC115679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK132588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK143245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK161226.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 BC115678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 BC115679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK132588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P347764 AK143245.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08521092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08521055 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08521081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161918 - -
          Whole Mouse Genome G4122A A_52_P347764 AK161226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 BC115678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 BC115679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 AK132588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 AK143245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 AK161226.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 BC115678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 BC115679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 AK132588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P347764 AK143245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470519 AK039092.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167024 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932516 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12932515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30574375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12755633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12932520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12572846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12538336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12538339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12555162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30574431 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438187 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438182 - -
          Affymetrix HT MG-430 PM Array Plate 1430714_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438178 - -
          Mouse Genome 430 2.0 Array 1430714_at - -
          Mouse Genome 430 2.0 Array 1430714_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600010523.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010523.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010523.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010523.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010523.mm.1 NM_029053.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010522.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010522.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010522.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010522.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010522.mm.1 NM_029053.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010518.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010518.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010518.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010518.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010518.mm.1 NM_029053.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010517.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010517.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010517.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010517.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010517.mm.1 NM_029053.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010520.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010520.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010520.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010520.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010520.mm.1 NM_029053.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010521.mm.1 TC1600001293.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010521.mm.1 NM_001145435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010521.mm.1 NM_001145435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010521.mm.1 NM_029053.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010521.mm.1 NM_029053.3 Refseq
          Mouse Expression Set 430 1430714_at - -
          Mouse Expression Set 430 1430714_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430714_at - -
          Affymetrix HT Mouse Genome 430B Array 1430714_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 748490 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103830575 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238803 - -
          Illumina Sentrix mouse-6.v1 bead array 103830575 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103830575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3006143 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238803 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3006143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0