Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01345762_g1
          See other B230119M05RIK GE Assays ›
          Gene Symbol:
          B230119M05Rik
          Gene Name:
          RIKEN cDNA B230119M05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 134684562 - 134686462 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01345762_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230119M05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045454.1 2 - 3 116 83
          GenBank mRNA AK020983.1 - 2 - 3 117 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY727074.1 - 2 - 3 117 Chr. X - 134684869 - 134686462 [-] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream armadillo repeat containing,
          X-linked 4
          Armcx4 AI448196,
          Gm195
          Anti-sense Chr. X - 134685651 - 134697775 on NCBI Build GRCm38
          Downstream armadillo repeat containing,
          X-linked 1
          Armcx1 3010033I09Rik,
          ALEX1
          32.2kb Anti-sense Chr. X - 134717912 - 134721917 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002997.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604809 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143105 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143105 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111266 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15003563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14644712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15052282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15003561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15144618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32673070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00018703.mm.1 TC0X00002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018703.mm.1 NR_045454.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018703.mm.1 NR_045454.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018702.mm.1 TC0X00002997.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018702.mm.1 NR_045454.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018702.mm.1 NR_045454.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 587641 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0