Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01347645_m1
          See other FAM47E GE Assays ›
          Gene Symbol:
          Fam47e
          Gene Name:
          family with sequence similarity 47, member E
          Gene Aliases:
          C730043O17, Gm1381
          Chromosome Location:
          Chr.5: 92554951 - 92591285 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01347645_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam47e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033478.2 NP_001028650.2 4 - 5 500 58
          GenBank mRNA AK144479.1 - 4 - 5 500 58
          BE650192.1 - 4 - 5 404 58

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK144479.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134265 BE650192.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134265 NM_001170571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134265 NM_001170572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134265 AA244788.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P134265 AK144479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960366 DV648863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960366 NM_001170571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960366 NM_001170572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960366 AK050399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960366 AK144479.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 362655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130487 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960366 DV648863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960366 NM_001170571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960366 NM_001170572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960366 AK050399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960366 AK144479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134265 BE650192.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134265 NM_001170571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134265 NM_001170572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134265 AA244788.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P134265 AK144479.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05656508 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05656518 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05656529 - -
          Whole Mouse Genome G4122A A_52_P859747 NM_001170571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P859747 NM_001170572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P859747 AK050399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P859747 NM_001033478.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05656508 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04134367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04273270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04265805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04134364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04265799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04265801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04746577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04134363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04978736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04977382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04352065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04795689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04368479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04408352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04467631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894891 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523251 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523253 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523252 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523247 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523250 - -
          Affymetrix HT MG-430 PM Array Plate 1455383_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523245 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 462781 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2990669 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2990669 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0