Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348186_m1
          See other PDE1C GE Assays ›
          Gene Symbol:
          Pde1c
          Gene Name:
          phosphodiesterase 1C
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 56069798 - 56652516 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348186_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025568.2 NP_001020739.1 5 - 6 1038 66
          NM_001159952.1 NP_001153424.1 4 - 5 1011 66
          NM_001159953.1 NP_001153425.1 5 - 6 1038 66
          NM_001159955.1 NP_001153427.1 6 - 7 600 66
          NM_001159956.1 NP_001153428.1 4 - 5 457 66
          NM_001159957.1 NP_001153429.1 5 - 6 484 66
          NM_001159960.1 NP_001153432.1 6 - 7 912 66
          NM_011054.4 NP_035184.1 6 - 7 1015 66
          GenBank mRNA AK014887.1 - 4 - 5 428 66
          AK015375.1 - 6 - 7 569 66
          AK029531.1 - 5 - 6 482 66
          AK030423.1 - 5 - 6 1040 66
          AK050499.1 - 6 - 7 913 66
          AK077123.1 - 4 - 5 406 66
          AK082180.1 - 5 - 6 1025 66
          AK082497.1 - 5 - 6 823 66
          AK133243.1 - 6 - 7 584 66
          AK139923.1 - 4 - 5 985 66
          BB614593.1 - 4 - 5 458 66
          BC032277.1 - 5 - 6 967 5400689 66
          L76944.1 - 5 - 6 668 66
          L76946.1 - 6 - 7 1014 66
          L76947.1 - 5 - 6 501 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002331.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pde1c

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP catabolic process
          signal transduction
          sensory perception of smell
          cGMP catabolic process
          negative regulation of insulin secretion
          response to calcium ion
          3',5'-cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
          calmodulin binding
          phosphoric diester hydrolase activity
          hydrolase activity
          cAMP binding
          metal ion binding
          calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 TC0600002331.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001025568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001025568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159952.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159952.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_001159960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_011054.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018228.mm.1 NM_011054.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 TC0600002331.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001025568.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001025568.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159952.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159952.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159953.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159953.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159955.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159955.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159956.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159956.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159957.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159957.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_001159960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_011054.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600018229.mm.1 NM_011054.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0