Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400032103.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032103.mm.1 |
NM_019701.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032103.mm.1 |
NM_019701.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032106.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032106.mm.1 |
NM_019701.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032106.mm.1 |
NM_019701.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032107.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032107.mm.1 |
NM_019701.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032107.mm.1 |
NM_019701.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032101.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032104.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032104.mm.1 |
NM_019701.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032104.mm.1 |
NM_019701.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032102.mm.1 |
TC0400003881.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400032102.mm.1 |
NM_019701.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400032102.mm.1 |
NM_019701.2 |
Refseq |