Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348848_m1
          See other CASP9 GE Assays ›
          Gene Symbol:
          Casp9
          Gene Name:
          caspase 9
          Gene Aliases:
          AI115399, APAF-3, AW493809, CASP-9, Caspase-9, ICE-LAP6, Mch6
          Chromosome Location:
          Chr.4: 141793612 - 141815978 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348848_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Casp9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277932.1 NP_001264861.1 5 - 6 1082 59
          NM_015733.5 NP_056548.2 5 - 6 1082 59
          GenBank mRNA AB019600.1 - 5 - 6 839 59
          AB019601.1 - 5 - 6 839 59
          AK085095.1 - 5 - 6 871 59
          AK085152.1 - 5 - 6 838 59
          AK154495.1 - 5 - 6 852 59
          AK156500.1 - 5 - 6 985 59
          AK162863.1 - 5 - 6 862 59
          BC056372.1 - 5 - 6 864 6834067 59
          BC056447.1 - 5 - 6 1082 5721188 59
          CT010400.1 - 5 - 6 839 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001801.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Casp9

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular response to DNA damage stimulus
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to UV
          response to organic cyclic compound
          cellular response to UV
          signal transduction in response to DNA damage
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of neuron apoptotic process
          cellular response to dexamethasone stimulus
          regulation of response to DNA damage stimulus
          cysteine-type endopeptidase activity
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          SH3 domain binding
          protein kinase binding
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400014956.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014956.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014956.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014955.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014955.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014955.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014953.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014953.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014953.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014954.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014954.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014954.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014957.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014957.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014957.mm.1 NM_015733.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014959.mm.1 TC0400001801.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014959.mm.1 NM_001277932.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014959.mm.1 NM_015733.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0