Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10572303 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10572304 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10572307 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10572306 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10572302 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10572305 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1429079_PM_a_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1456267_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10572301 |
- |
- |
Mouse Genome 430 2.0 Array |
1429079_PM_a_at |
- |
- |
Mouse Genome 430 2.0 Array |
1429079_a_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
72368_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
72368_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800005067.mm.1 |
TC0800003226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005067.mm.1 |
NM_001145552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005070.mm.1 |
TC0800003226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005070.mm.1 |
NM_001145552.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800005071.mm.1 |
TC0800003226.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800005071.mm.1 |
NM_001145552.2 |
Refseq |
Mouse Expression Set 430 |
1429079_a_at |
- |
- |
Mouse Expression Set 430 |
1429079_PM_a_at |
- |
- |