Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004062.2 | NP_001004062.1 | 1 - 2 | 139 | 78 | |
GenBank mRNA | AK129173.1 | - | 1 - 2 | 136 | 78 | |
BC056137.1 | - | 1 - 2 | 145 | 78 | ||
BC068139.1 | - | 1 - 2 | 145 | 78 | ||
BC080308.1 | - | 1 - 2 | 139 | 5719562 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |