Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01349301_m1
          See other CLTA GE Assays ›
          Gene Symbol:
          Clta
          Gene Name:
          clathrin, light polypeptide (Lca)
          Gene Aliases:
          AV026556, Lca
          Chromosome Location:
          Chr.4: 44003816 - 44032848 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01349301_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clta
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080384.1 NP_001073853.1 1 - 2 366 76
          NM_001080385.1 NP_001073854.1 1 - 2 366 76
          NM_001290470.1 NP_001277399.1 1 - 2 359 76
          NM_016760.2 NP_058040.2 1 - 2 366 76
          GenBank mRNA AK146965.1 - 2 - 3 999 76
          AK149761.1 - 1 - 2 332 76
          AK150148.1 - 1 - 2 332 76
          AK152397.1 - 1 - 2 336 76
          AK168130.1 - 1 - 2 360 76
          BC057660.1 - 1 - 2 326 3154277 76
          BU847112.1 - 1 - 2 373 76
          CA322069.1 - 1 - 2 329 76
          CK620776.1 - 1 - 2 350 76
          CX243404.1 - 1 - 2 318 76
          U91848.1 - 1 - 2 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000460.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clta

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle-mediated transport
          clathrin-dependent endocytosis
          structural molecule activity
          clathrin heavy chain binding
          peptide binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 TC0400000460.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080384.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080384.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080386.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_001080386.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_016760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003235.mm.1 NM_016760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 TC0400000460.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080384.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080384.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080386.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_001080386.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_016760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003237.mm.1 NM_016760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 TC0400000460.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080384.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080384.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080385.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080385.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080386.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_001080386.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_016760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400003232.mm.1 NM_016760.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 335422 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc