Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028878.3 | NP_083154.1 | 5 - 6 | 841 | 67 | |
GenBank mRNA | AJ633679.1 | - | 5 - 6 | 850 | 67 | |
AK014544.1 | - | 5 - 6 | 842 | 67 | ||
AK028784.1 | - | 5 - 6 | 842 | 67 | ||
AK162399.1 | - | 4 - 5 | 586 | 67 | ||
BC107411.1 | - | 5 - 6 | 842 | 40059042 | 67 | |
BC107412.2 | - | 5 - 6 | 846 | 40059050 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |