Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352645_g1
          See other PRICKLE3 GE Assays ›
          Gene Symbol:
          Prickle3
          Gene Name:
          prickle homolog 3 (Drosophila)
          Gene Aliases:
          Lmo6, Sfc16
          Chromosome Location:
          Chr.X: 7657379 - 7668186 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352645_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prickle3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290624.1 NP_001277553.1 8 - 9 1337 61
          GenBank mRNA AK037217.1 - 8 - 9 1300 61
          AK083704.1 - 7 - 8 1304 61
          BC023671.1 - 8 - 9 1333 61
          BC032173.1 - 5 - 6 1658 61
          BC049258.1 - 8 - 9 1279 5250488 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000061.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 BC032173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 BC049258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 AK083704.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 NM_175097.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 AK037217.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 BC032173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 BC049258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 AK083704.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 NM_175097.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P133792 AK037217.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 963762 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107040 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 BC032173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 BC049258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 AK083704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 NM_175097.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 AK037217.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 BC032173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 BC049258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 AK083704.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 NM_175097.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P133792 AK037217.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32239516 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107040 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09226452 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136421 - -
          Whole Mouse Genome G4122A A_51_P133792 BC032173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 BC049258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 AK083704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 NM_175097.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 AK037217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 BC032173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 BC049258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 AK083704.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 NM_175097.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133792 AK037217.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1048 AK079845.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131046 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107040 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878181 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878201 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878209 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878207 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14878205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32239616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14643977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14714456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14763943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14963605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14878177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14864034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14643972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14963601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14963620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14746048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14878176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14641340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14963611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32239499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10411360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598374 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603347 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603348 - -
          Affymetrix HT MG-430 PM Array Plate 1439296_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598369 - -
          Mouse Genome 430 2.0 Array 1439296_at - -
          Mouse Genome 430 2.0 Array 1439296_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00000326.mm.1 TC0X00000061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000327.mm.1 TC0X00000061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000328.mm.1 TC0X00000061.mm.1 MTA TC
          Mouse Expression Set 430 1439296_PM_at - -
          Mouse Expression Set 430 1439296_at - -
          Mouse Genome 430A 2.0 Array 1439296_at - -
          Mouse Genome 430A 2.0 Array 1439296_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439296_at - -
          Affymetrix HT Mouse Genome 430A Array 1439296_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 928427 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4210072 - -
          Illumina mouse-6 v1.0 expression beadchip 4210072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602858 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739398 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602858 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2739398 - -
          mouse-6 v1.1 (Illumina) scl55078.9.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl55078.9.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739398 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739398 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl55078.9.1_18-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0