Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01354317_g1
          See other CXCL5 GE Assays ›
          Gene Symbol:
          Cxcl5
          Gene Name:
          chemokine (C-X-C motif) ligand 5
          Gene Aliases:
          AMCF-II, ENA-78, GCP-2, LIX, Scyb5, Scyb6
          Chromosome Location:
          Chr.5: 90759298 - 90761625 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01354317_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009141.3 NP_033167.2 3 - 4 481 79
          GenBank mRNA AK044397.1 - 1 - 2 690 79
          BC024392.1 - 3 - 4 393 4976854 79
          BQ829929.1 - 3 - 4 399 79
          U27267.1 - 3 - 4 401 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000841.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl5

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte homeostasis
          cytokine production
          positive regulation of leukocyte chemotaxis
          chemotaxis
          inflammatory response
          immune response
          G protein-coupled receptor signaling pathway
          positive regulation of neuron projection development
          animal organ regeneration
          response to lipopolysaccharide
          regulation of chemokine production
          regulation of cell population proliferation
          positive regulation of receptor signaling pathway via JAK-STAT
          cell chemotaxis
          chemokine-mediated signaling pathway
          regulation of neutrophil mediated killing of gram-negative bacterium
          cytokine activity
          chemokine activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990032 NM_009141.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990032 AK044397.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 593502 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113911 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990032 NM_009141.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990032 AK044397.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295432 U27267.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295432 BC024392.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295432 NM_009141.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295432 AK044397.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P295432 AK156787.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23884999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885033 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113911 - -
          Whole Mouse Genome G4122A A_52_P295432 U27267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295432 BC024392.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295432 NM_009141.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295432 AK044397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295432 AK156787.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04978351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04978358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04217790 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04978347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04978356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04978355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23884968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04654257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04887078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04217788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04978358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04887076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04311773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23884978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885026 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523126 - -
          Affymetrix HT MG-430 PM Array Plate 1419728_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523120 - -
          Mouse Genome 430 2.0 Array 1419728_at - -
          Mouse Genome 430 2.0 Array 1419728_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20311_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20311_at - -
          Affymetrix Murine Genome 6500A Array Msa16130_at - -
          Affymetrix Mouse Clariom S Array PSR0500007408.mm.1 TC0500000841.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007408.mm.1 NM_009141.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007410.mm.1 TC0500000841.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007410.mm.1 NM_009141.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007409.mm.1 TC0500000841.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007409.mm.1 NM_009141.3 Refseq
          Mouse Expression Set 430 1419728_PM_at - -
          Mouse Expression Set 430 1419728_at - -
          Murine Genome U74v2 Set 98772_PM_at - -
          Murine Genome U74v2 Set 98772_at - -
          Mouse Genome 430A 2.0 Array 1419728_at - -
          Mouse Genome 430A 2.0 Array 1419728_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419728_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419728_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395074 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6370333 - -
          Illumina mouse-6 v1.0 expression beadchip 6370333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230278 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230278 - -
          mouse-6 v1.1 (Illumina) scl27604.2_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27604.2_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230278 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27604.2_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0