Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01519304_g1
          See other HDAC1 GE Assays ›
          Gene Symbol:
          Hdac1
          Gene Name:
          histone deacetylase 1
          Gene Aliases:
          HD1, Hdac1-ps, MommeD5, RPD3
          Chromosome Location:
          Chr.4: 129516104 - 129542646 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01519304_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hdac1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008228.2 NP_032254.1 1 - 2 63 70
          GenBank mRNA BC092070.1 - 1 - 2 58 4217199 70
          BC092285.1 - 1 - 2 63 70
          BC108371.1 - 1 - 2 64 4976514 70
          U80780.1 - 1 - 2 60 70
          X98207.1 - 1 - 2 63 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hdac1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          response to amphetamine
          chromatin organization
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          protein deacetylation
          endoderm development
          circadian rhythm
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          epidermal cell differentiation
          negative regulation of gene expression
          histone deacetylation
          hippocampus development
          neuron differentiation
          positive regulation of interleukin-1 production
          positive regulation of tumor necrosis factor production
          circadian regulation of gene expression
          cellular response to oxidative stress
          odontogenesis of dentin-containing tooth
          positive regulation of tyrosine phosphorylation of STAT protein
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of neuron apoptotic process
          negative regulation by host of viral transcription
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of insulin secretion
          rhythmic process
          positive regulation of oligodendrocyte differentiation
          negative regulation of androgen receptor signaling pathway
          hair follicle placode formation
          eyelid development in camera-type eye
          fungiform papilla formation
          histone H3 deacetylation
          histone H4 deacetylation
          negative regulation of canonical Wnt signaling pathway
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          positive regulation of type B pancreatic cell apoptotic process
          negative regulation of peptidyl-lysine acetylation
          negative regulation of intrinsic apoptotic signaling pathway
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          transcription corepressor activity
          histone deacetylase activity
          protein binding
          transcription factor binding
          hydrolase activity
          deacetylase activity
          enzyme binding
          histone deacetylase activity (H3-K14 specific)
          nucleosomal DNA binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase activity (H3-K9 specific)
          protein deacetylase activity
          histone deacetylase activity (H4-K16 specific)
          Krueppel-associated box domain binding
          histone deacetylase binding
          protein-containing complex binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          protein N-terminus binding
          NF-kappaB binding
          RNA polymerase II-specific DNA-binding transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K18 specific)
          DNA-binding transcription factor binding

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892921 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0