Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01545339_m1
          See other A530058N18RIK GE Assays ›
          Gene Symbol:
          A530058N18Rik
          Gene Name:
          RIKEN cDNA A530058N18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 114013565 - 114032292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01545339_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A530058N18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028423.1 2 - 3 376 81
          GenBank mRNA AK045761.1 - 2 - 3 232 81
          AK045885.1 - 2 - 3 181 81
          AK046235.1 - 2 - 3 206 81

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK045885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK046235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK045761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK043120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 NR_028423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK045885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK046235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK045761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 AK043120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373575 NR_028423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633349 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK045885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK046235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK045761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK043120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 NR_028423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK045885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK046235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK045761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 AK043120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373575 NR_028423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK045885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK046235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK045761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK043120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 NR_028423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK045885.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK046235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK045761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 AK043120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373575 NR_028423.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P939223 AK044743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113522 AK082668.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P01726639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01227534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01227529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01227537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01726625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01883789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01726627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21537508 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1443313_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447058_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438746_PM_at - -
          Mouse Genome 430 2.0 Array 1443470_at - -
          Mouse Genome 430 2.0 Array 1438746_at - -
          Mouse Genome 430 2.0 Array 1443313_at - -
          Mouse Genome 430 2.0 Array 1447058_at - -
          Mouse Genome 430 2.0 Array 1447058_PM_at - -
          Mouse Genome 430 2.0 Array 1443313_PM_at - -
          Mouse Genome 430 2.0 Array 1438746_PM_at - -
          Mouse Genome 430 2.0 Array 1443470_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320846_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320846_PM_at - -
          Mouse Expression Set 430 1438746_at - -
          Mouse Expression Set 430 1438746_PM_at - -
          Mouse Expression Set 430 1443470_PM_at - -
          Mouse Expression Set 430 1443313_PM_at - -
          Mouse Expression Set 430 1447058_PM_at - -
          Mouse Expression Set 430 1447058_at - -
          Mouse Expression Set 430 1443470_at - -
          Mouse Expression Set 430 1443313_at - -
          Murine Genome U74v2 Set 136454_PM_at - -
          Murine Genome U74v2 Set 136454_at - -
          Mouse Genome 430A 2.0 Array 1438746_PM_at - -
          Mouse Genome 430A 2.0 Array 1438746_at - -
          Affymetrix HT Mouse Genome 430A Array 1438746_at - -
          Affymetrix HT Mouse Genome 430A Array 1438746_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443470_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443470_at - -
          Affymetrix HT Mouse Genome 430B Array 1443313_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447058_at - -
          Affymetrix HT Mouse Genome 430B Array 1447058_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443313_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 773033 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101580670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0