Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01598638_g1
          See other CCDC115 GE Assays ›
          Gene Symbol:
          Ccdc115
          Gene Name:
          coiled-coil domain containing 115
          Gene Aliases:
          2310061I09Rik, Ccp1
          Chromosome Location:
          Chr.1: 34436670 - 34439672 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01598638_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027159.2 NP_081435.1 4 - 5 770 76
          GenBank mRNA AK010002.1 - 4 - 5 464 76
          AK151074.1 - 1 - 2 976 76
          AK154179.1 - 4 - 5 470 76
          AK162237.1 - 4 - 5 383 76
          BC019430.1 - 4 - 5 771 3491825 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002151.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc115

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138845 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138849 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1954081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97427 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1954081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20138849 - -
          Whole Mouse Genome G4122A A_52_P319149 AK162237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319149 AK010002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319149 AK151074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319149 AK154179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319149 NM_027159.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00556205 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00556212 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00556202 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20138845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20138848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00726694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00726690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00556202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00160140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00516985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00726689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00631687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00414699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20138837 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353785 - -
          Affymetrix HT MG-430 PM Array Plate 1451265_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10353783 - -
          Mouse Genome 430 2.0 Array 1451265_PM_at - -
          Mouse Genome 430 2.0 Array 1451265_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69668_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69668_at - -
          Affymetrix Mouse Clariom S Array PSR0100017251.mm.1 TC0100002151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017251.mm.1 NM_027159.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017251.mm.1 NM_027159.2 Refseq
          Mouse Expression Set 430 1451265_at - -
          Mouse Expression Set 430 1451265_PM_at - -
          Murine Genome U74v2 Set 101404_at - -
          Murine Genome U74v2 Set 101404_PM_at - -
          Murine Genome U74v2 Set 171214_PM_at - -
          Murine Genome U74v2 Set 171214_at - -
          Mouse Genome 430A 2.0 Array 1451265_at - -
          Mouse Genome 430A 2.0 Array 1451265_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451265_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451265_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 810326 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2672035 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243032 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215353 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2672035 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215353 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243032 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0