Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01605834_g1
          See other LAMTOR5 GE Assays ›
          Gene Symbol:
          Lamtor5
          Gene Name:
          late endosomal/lysosomal adaptor, MAPK and MTOR activator 5
          Gene Aliases:
          1110003H18Rik, Hbxip, XIP
          Chromosome Location:
          Chr.3: 107278858 - 107284082 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01605834_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamtor5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026774.2 NP_081050.2 3 - 4 429 68
          GenBank mRNA AK003358.1 - 3 - 4 269 68
          AK146289.1 - 3 - 4 289 68
          BC028547.1 - 3 - 4 259 1513442 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001116.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell size
          viral genome replication
          positive regulation of TOR signaling
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of GTPase activity
          protein localization to lysosome
          cellular response to amino acid stimulus
          guanyl-nucleotide exchange factor activity
          molecular adaptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405342 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405343 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088416 AK003358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088416 AK146289.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088416 BC028547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088416 NM_026774.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45861 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140645 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088416 AK003358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088416 AK146289.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088416 BC028547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088416 NM_026774.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140645 - -
          Whole Mouse Genome G4122A A_51_P205865 AK003358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205865 AK146289.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205865 BC028547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P205865 NM_026774.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405342 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405343 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405341 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22405344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22405369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02802351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02293418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02745866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03130123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02293419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03123378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02989739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03104476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02653614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02397941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02753884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22405299 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10495204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495203 - -
          Affymetrix HT MG-430 PM Array Plate 1436152_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1415759_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10607585 - -
          Affymetrix Mouse Gene 1.1 ST Array 10495201 - -
          Mouse Genome 430 2.0 Array 1436152_a_at - -
          Mouse Genome 430 2.0 Array 1445400_at - -
          Mouse Genome 430 2.0 Array 1436152_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415759_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415759_a_at - -
          Mouse Genome 430 2.0 Array 1445400_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68576_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68576_at - -
          Affymetrix Mouse Clariom S Array PSR0300009085.mm.1 TC0300001116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300009085.mm.1 NM_026774.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009085.mm.1 NM_026774.2 Refseq
          Mouse Expression Set 430 1436152_PM_a_at - -
          Mouse Expression Set 430 1415759_PM_a_at - -
          Mouse Expression Set 430 1415759_a_at - -
          Mouse Expression Set 430 1436152_a_at - -
          Mouse Expression Set 430 1415759_a_at - -
          Mouse Expression Set 430 1415759_PM_a_at - -
          Murine Genome U74v2 Set 98037_PM_at - -
          Murine Genome U74v2 Set 98037_at - -
          Mouse Genome 430A 2.0 Array 1415759_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1436152_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1436152_a_at - -
          Mouse Genome 430A 2.0 Array 1415759_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1436152_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415759_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415759_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1436152_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415759_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415759_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 609925 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2030279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251708 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251708 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline