Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01612121_gH
          See other RPS3A1 GE Assays ›
          Gene Symbol:
          Rps3a1
          Gene Name:
          ribosomal protein S3A1
          Gene Aliases:
          Rps3a
          Chromosome Location:
          Chr.3: 86137940 - 86142668 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01612121_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016959.4 NP_058655.3 2 156
          GenBank mRNA AK003158.1 - 2 156
          AK010605.1 - 2 156
          AK012345.1 - 2 156
          AK050610.1 - 2 156
          AK088113.1 - 2 156
          AK135318.1 - 2 156
          AK146304.1 - 2 156
          AK148513.1 - 2 156
          AK150441.1 - 2 156
          AK150796.1 - 2 156
          AK150814.1 - 2 156
          AK151982.1 - 2 156
          AK153480.1 - 2 156
          AK159777.1 - 2 156
          AK167512.1 - 2 156
          AK167562.1 - 2 156
          AK168240.1 - 2 156
          AL118161.1 - 2 156
          BC039659.1 - 2 5025347 156
          BC081451.1 - 2 6824188 156
          BC084675.1 - 2 5319512 156
          BC106115.1 - 2 2599128 156

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002263.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK003158.1, AK010605.1, AK012345.1, AK050610.1, AK088113.1, AK135318.1, AK146304.1, AK148513.1, AK150441.1, AK150796.1, AK150814.1, AK151982.1, AK153480.1, AK159777.1, AK167512.1, AK167562.1, AK168240.1, BC039659.1, BC081451.1, BC084675.1, BC106115.1, NM_016959.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps3a1

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          cell differentiation
          structural constituent of ribosome
          protein binding
          translation initiation factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300017039.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017039.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017039.mm.1 NM_016959.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017037.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017037.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017037.mm.1 NM_016959.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017038.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017038.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017038.mm.1 NM_016959.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017040.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017040.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017040.mm.1 NM_016959.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 898494 - -
          ABI Mouse Genome Survey Microarray 901185 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0