Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01613463_s1
          See other HIST2H2AB GE Assays ›
          Gene Symbol:
          Hist2h2ab
          Gene Name:
          histone cluster 2, H2ab
          Gene Aliases:
          EG621893, H2a-613a
          Chromosome Location:
          Chr.3: 96219916 - 96220353 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01613463_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist2h2ab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178213.4 NP_835585.3 1 158 167

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000929.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Process(es)

          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348221 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114176 NM_178213.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114176 NM_178213.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22348227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02752604 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22348222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22348214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02872678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02386738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02752589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22348205 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494387 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494386 - -
          Affymetrix Mouse Clariom S Array PSR0300007443.mm.1 TC0300000929.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007443.mm.1 NM_178213.4 Refseq
          Murine Genome U74v2 Set 102557_PM_at - -
          Murine Genome U74v2 Set 102557_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 715000 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102260563 - -
          Illumina mouse-6 v1.0 expression beadchip 102260563 - -
          mouse-6 v1.1 (Illumina) GI_37059773-S - -
          Illumina Sentrix mouse-6.v1 bead array 102260563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102260563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2785454 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2785454 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254276 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0