Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01616065_g1
          See other RPS3A1 GE Assays ›
          Gene Symbol:
          Rps3a1
          Gene Name:
          ribosomal protein S3A1
          Gene Aliases:
          Rps3a
          Chromosome Location:
          Chr.3: 86137940 - 86142668 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01616065_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016959.4 NP_058655.3 1 - 2 166 143
          GenBank mRNA AL118161.1 - 1 - 2 171 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002263.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the NM_016959.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rps3a1

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          cell differentiation
          structural constituent of ribosome
          protein binding
          translation initiation factor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301213 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2056190 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136385 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 M88335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 BC084675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 AK146304.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 M88335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 BC084675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292357 AK146304.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961943 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174185 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2056190 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136385 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 M88335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 BC084675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 AK146304.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 M88335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 BC084675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292357 AK146304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 M88335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 BC084675.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 AK146304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 M88335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 BC084675.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292357 AK146304.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P49886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22301234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174185 - -
          Whole Mouse Genome G4122A A_51_P292357 M88335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292357 BC084675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292357 AK146304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292357 M88335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292357 BC084675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292357 AK146304.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P49886 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123074 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105515 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02568720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02568721 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301213 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22301215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22301180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02655188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02816716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02992440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02446947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02372073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02948936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02568434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02568722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02460069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03200594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22301177 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421909 - -
          Affymetrix Mouse Gene 1.1 ST Array 10499121 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407120 - -
          Affymetrix Mouse Gene 1.1 ST Array 10421908 - -
          Mouse Genome 430 2.0 Array 1422475_PM_a_at - -
          Mouse Genome 430 2.0 Array 1422475_a_at - -
          Affymetrix Mouse Clariom S Array PSR0300017041.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017041.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017041.mm.1 NM_016959.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017046.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017046.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017046.mm.1 NM_016959.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017042.mm.1 TC0300002263.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017042.mm.1 NM_016959.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017042.mm.1 NM_016959.4 Refseq
          Mouse Expression Set 430 1422475_a_at - -
          Mouse Expression Set 430 1422475_PM_a_at - -
          Murine Genome U74v2 Set 101664_at - -
          Murine Genome U74v2 Set 101664_PM_at - -
          Mouse Genome 430A 2.0 Array 1422475_a_at - -
          Mouse Genome 430A 2.0 Array 1422475_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 903437 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470086 - -
          Illumina mouse-6 v1.0 expression beadchip 2470086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227285 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227285 - -
          mouse-6 v1.1 (Illumina) scl020091.3_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020091.3_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2733330 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2733330 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020091.3_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0