Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01621044_g1
          See other CYCS GE Assays ›
          Gene Symbol:
          Cycs
          Gene Name:
          cytochrome c, somatic
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 50562563 - 50566474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01621044_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cycs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007808.4 NP_031834.1 3 1060 144
          GenBank mRNA AK002732.1 - 3 1062 144
          AK143887.1 - 3 1061 144
          BC034363.1 - 3 1044 2650742 144
          BC094054.1 - 3 1049 2865963 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002253.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cycs

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial electron transport, ubiquinol to cytochrome c
          mitochondrial electron transport, cytochrome c to oxygen
          apoptotic DNA fragmentation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
          hydrogen peroxide metabolic process
          ATP synthesis coupled electron transport
          electron transfer activity
          heme binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600017702.mm.1 TC0600002253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017702.mm.1 NM_007808.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017702.mm.1 NM_007808.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017700.mm.1 TC0600002253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017700.mm.1 NM_007808.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017700.mm.1 NM_007808.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017699.mm.1 TC0600002253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017699.mm.1 NM_007808.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017699.mm.1 NM_007808.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017701.mm.1 TC0600002253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017701.mm.1 NM_007808.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017701.mm.1 NM_007808.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017704.mm.1 TC0600002253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017704.mm.1 NM_007808.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017704.mm.1 NM_007808.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894005 - -
          ABI Mouse Genome Survey Microarray 900401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0