Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01623171_g1
          See other RSL24D1 GE Assays ›
          Gene Symbol:
          Rsl24d1
          Gene Name:
          ribosomal L24 domain containing 1
          Gene Aliases:
          2410159K22Rik
          Chromosome Location:
          Chr.9: 73113469 - 73123333 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01623171_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsl24d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198609.2 NP_941011.1 6 863 168
          GenBank mRNA AK153492.1 - 6 874 168
          AK164856.1 - 6 860 169
          BC089481.1 - 6 860 30254073 168

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003328.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsl24d1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ribosome biogenesis
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900028038.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900028038.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900028038.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007719.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007719.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007719.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007711.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007711.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007711.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007715.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007715.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007715.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007716.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007716.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007716.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007720.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007720.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007720.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007712.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007712.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007712.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007718.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007718.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007718.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007708.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007708.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007708.mm.1 NM_198609.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007717.mm.1 TC0900003328.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007717.mm.1 NM_198609.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007717.mm.1 NM_198609.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 694977 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0