Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701133_g1
          See other PRR9 GE Assays ›
          Gene Symbol:
          Prr9
          Gene Name:
          proline rich 9
          Gene Aliases:
          A030004J04Rik
          Chromosome Location:
          Chr.3: 92122204 - 92123947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701133_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prr9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175424.3 NP_780633.1 1 - 2 43 86
          GenBank mRNA AK079463.1 - 1 - 2 44 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002384.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK079463.1, NM_175424.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prr9

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009732 NM_175424.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009732 AK079463.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530169 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156432 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009732 NM_175424.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009732 AK079463.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22329823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156432 - -
          Whole Mouse Genome G4122A A_52_P160408 NM_175424.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P160408 AK079463.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110946 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02390058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02390048 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02390061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02390052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02841093 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02390053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22329729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02447304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03199187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03028245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02447293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02447310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02320580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02817649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03019219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02841087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02390061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22329771 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499874 - -
          Affymetrix HT MG-430 PM Array Plate 1444061_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499873 - -
          Mouse Genome 430 2.0 Array 1444061_PM_at - -
          Mouse Genome 430 2.0 Array 1444061_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109314_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109314_at - -
          Affymetrix Mouse Clariom S Array PSR0300018548.mm.1 TC0300002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018548.mm.1 NM_175424.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018548.mm.1 NM_175424.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018547.mm.1 TC0300002384.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018547.mm.1 NM_175424.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018547.mm.1 NM_175424.3 Refseq
          Mouse Expression Set 430 1444061_PM_at - -
          Mouse Expression Set 430 1444061_at - -
          Affymetrix HT Mouse Genome 430B Array 1444061_at - -
          Affymetrix HT Mouse Genome 430B Array 1444061_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 677216 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130725 - -
          Illumina mouse-6 v1.0 expression beadchip 6130725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130725 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623605 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623605 - -
          mouse-6 v1.1 (Illumina) scl21904.2.1_253-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130725 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21904.2.1_253-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623605 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0