Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701806_m1
          See other KRT9 GE Assays ›
          Gene Symbol:
          Krt9
          Gene Name:
          keratin 9
          Gene Aliases:
          EPPK, K9, Krt1-9
          Chromosome Location:
          Chr.11: 100186781 - 100193875 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701806_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201255.2 NP_957707.2 7 - 8 2259 74
          GenBank mRNA AY497550.1 - 7 - 8 2259 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003759.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt9

          Gene Ontology Categories:

          Function(s) Process(es)

          spermatogenesis
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060667 NM_201255.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060667 AY497550.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164634 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136627 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060667 NM_201255.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060667 AY497550.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P307860 NM_201255.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P307860 AY497550.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28066682 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07463113 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136627 - -
          Whole Mouse Genome G4122A A_52_P307860 NM_201255.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P307860 AY497550.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139256 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09015128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09015125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09015123 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09015126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09015122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28066718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09163107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09310091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09572921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09445398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09445399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09527949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09033171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09382837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09453876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09680462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09680463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08985393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09033174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28066703 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10391050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391045 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391044 - -
          Affymetrix Mouse Gene 1.1 ST Array 10391043 - -
          Affymetrix Mouse Clariom S Array PSR1100034493.mm.1 TC1100003759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100034493.mm.1 NM_201255.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034493.mm.1 NM_201255.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6510593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006510593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2748165 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2748165 - -
          mouse-6 v1.1 (Illumina) scl0107656.2_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 6510593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0107656.2_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6510593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2748165 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748165 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0107656.2_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0