Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702876_mH
          See other DNAJB6 GE Assays ›
          Gene Symbol:
          Dnajb6
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 6
          Gene Aliases:
          Mrj, mDj4
          Chromosome Location:
          Chr.5: 29735687 - 29786478 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702876_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajb6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037940.4 NP_001033029.1 1 - 2 217 140
          NM_001037941.3 NP_001033030.2 1 - 2 217 140
          NM_001127367.1 NP_001120839.1 1 - 2 217 140
          NM_011847.4 NP_035977.2 1 - 2 217 140
          GenBank mRNA AB028854.1 - 1 - 2 119 140
          AF035962.1 - 1 - 2 117 139
          AK005680.1 - 1 - 2 122 141
          AK032039.1 - 1 - 2 126 140
          AK133805.1 - 1 - 2 134 140
          AK134387.1 - 1 - 2 176 140
          AK144182.1 - 1 - 2 144 140
          AK151219.1 - 1 - 2 124 140
          AK152626.1 - 1 - 2 131 140
          AK169332.1 - 1 - 2 123 140
          BC083349.1 - 1 - 2 109 30541723 140
          BG800394.1 - 1 - 2 107 140
          CJ079100.1 - 1 - 2 217 140

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB028854.1, AF035962.1, AK005680.1, AK133805.1, AK134387.1, AK144182.1, AK151219.1, AK152626.1, AK169332.1, BC083349.1, NM_001037940.4, NM_001037941.3, NM_001127367.1, NM_011847.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajb6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          actin cytoskeleton organization
          extracellular matrix organization
          positive regulation of ATPase activity
          protein localization to nucleus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          intermediate filament organization
          negative regulation of transcription, DNA-templated
          chorio-allantoic fusion
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          chorion development
          chaperone-mediated protein folding
          negative regulation of inclusion body assembly
          ATPase activator activity
          DNA binding
          protein binding
          heat shock protein binding
          protein folding chaperone
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894464 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0