Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01716145_m1
          See other CMSS1 GE Assays ›
          Gene Symbol:
          Cmss1
          Gene Name:
          cms small ribosomal subunit 1
          Gene Aliases:
          1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629
          Chromosome Location:
          Chr.16: 57302000 - 57606867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01716145_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cmss1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003195.1 - 1 - 2 180 72
          AK080386.1 - 1 - 2 172 72
          AK136657.1 - 1 - 2 204 72
          BC038682.1 - 1 - 2 95 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY102771.1 - 1 - 2 140 Chr. 16 - 57561564 - 57606803 [-] on Build GRCm38 72
          GenBankmRNA BB682144.1 - 1 - 2 173 Chr. 16 - 57561441 - 57606837 [-] on Build GRCm38 72
          GenBankmRNA AI120191.1 - 1 - 2 101 Chr. 16 - 57561431 - 57606759 [-] on Build GRCm38 72
          GenBankmRNA BX512053.1 - 1 - 2 Chr. 16 - 57561433 - 57606770 [+] on Build GRCm38 72
          GenBankmRNA AA461767.1 - 1 - 2 Chr. 16 - 57561435 - 57606733 [+] on Build GRCm38 73
          GenBankmRNA BY721034.1 - 1 - 2 200 Chr. 16 - 57561392 - 57606864 [-] on Build GRCm38 72
          GenBankmRNA CB056860.1 - 1 - 2 165 Chr. 16 - 57561408 - 57606821 [-] on Build GRCm38 72
          GenBankmRNA BY102783.1 - 1 - 2 140 Chr. 16 - 57561563 - 57606803 [-] on Build GRCm38 72
          GenBankmRNA BY105473.1 - 1 - 2 183 Chr. 16 - 57561557 - 57606846 [-] on Build GRCm38 72
          GenBankmRNA BB639315.1 - 1 - 2 172 Chr. 16 - 57561475 - 57606836 [-] on Build GRCm38 72
          GenBankmRNA CA465091.1 - 1 - 2 144 Chr. 16 - 57561381 - 57606806 [-] on Build GRCm38 72
          GenBankmRNA BU924933.1 - 1 - 2 182 Chr. 16 - 57561431 - 57606846 [-] on Build GRCm38 72
          GenBankmRNA CA493059.1 - 208 Chr. - - [-] on Build 72
          GenBankmRNA BX634056.1 - 1 - 2 Chr. 16 - 57561433 - 57606782 [+] on Build GRCm38 72
          GenBankmRNA AA549307.1 - 1 - 2 117 Chr. 16 - 57561445 - 57606779 [-] on Build GRCm38 72
          GenBankmRNA AI047162.1 - 1 - 2 Chr. 16 - 57561458 - 57606772 [+] on Build GRCm38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream filamin A interacting protein 1-like Filip1l 4631422O05Rik,
          Doc1
          Sense Chr. 16 - 57353277 - 57572804 on NCBI Build GRCm38
          Downstream collagen,
          type VIII,
          alpha 1
          Col8a1 Col8a-1 17.5kb Anti-sense Chr. 16 - 57624256 - 57754737 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0