Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01717319_s1
          See other 4930528A17RIK GE Assays ›
          Gene Symbol:
          4930528A17Rik
          Gene Name:
          RIKEN cDNA 4930528A17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 21846327 - 21848448 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01717319_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930528A17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028384.1 1 958 82
          GenBank mRNA AK014838.1 - 1 960 82
          AK015922.1 - 1 880 82
          AK039347.1 - 1 942 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitin specific petidase 45 Usp45 3110003C05Rik,
          4930550B20Rik,
          AI843191,
          Gcap7
          -8.5kb Anti-sense Chr. 4 - 21767161 - 21837872 on NCBI Build GRCm38
          Downstream coenzyme Q3 homolog,
          methyltransferase (yeast)
          Coq3 4732433J24,
          C77934
          31.2kb Anti-sense Chr. 4 - 21879675 - 21912163 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002291.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064976 AK015922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064976 AK039347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064976 BC031206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064976 NR_028384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2064976 AK014838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK015922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK039347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 BC031206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 NR_028384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK014838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK015922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK039347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 BC031206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 NR_028384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P459259 AK014838.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208615 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155583 - -
          Whole Mouse Genome G4122A A_52_P459259 AK015922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 AK039347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 BC031206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 NR_028384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 AK014838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 AK015922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 AK039347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 BC031206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 NR_028384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P459259 AK014838.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22775165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22775169 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10511809 - -
          Affymetrix HT MG-430 PM Array Plate 1430616_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10511808 - -
          Mouse Genome 430 2.0 Array 1430616_at - -
          Mouse Genome 430 2.0 Array 1430616_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67735_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67735_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400018948.mm.1 TC0400002291.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400018948.mm.1 NR_028384.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400018948.mm.1 NR_028384.1 Refseq
          Mouse Expression Set 430 1430616_at - -
          Mouse Expression Set 430 1430616_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430616_at - -
          Affymetrix HT Mouse Genome 430B Array 1430616_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 563726 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102320497 - -
          MouseWG-6 v2 Expression BeadChip 106100358 - -
          MouseWG-6 v2 Expression BeadChip 104590358 - -
          Illumina mouse-6 v1.0 expression beadchip 106100358 - -
          Illumina mouse-6 v1.0 expression beadchip 104590358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231706 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231861 - -
          Illumina Sentrix mouse-6.v1 bead array 104590358 - -
          Illumina Sentrix mouse-6.v1 bead array 106100358 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106100358 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104590358 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2549714 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231706 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0