Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718070_m1
          See other A330094K24RIK GE Assays ›
          Gene Symbol:
          A330094K24Rik
          Gene Name:
          RIKEN cDNA A330094K24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718070_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330094K24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020726.1 - 1 - 2 229 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930465K10 gene 4930465K10Rik -14.2kb Anti-sense Chr. 18 - 77714184 - 77715445 on NCBI Build GRCm38
          Downstream HAUS augmin-like complex,
          subunit 1
          Haus1 BC024400,
          Ccdc5,
          HEI-C
          24.1kb Sense Chr. 18 - 77757567 - 77773847 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2351300 AK020726.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 752977 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102706 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102706 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131983 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13622626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14113546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13986812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31849554 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1460479_PM_at - -
          Mouse Genome 430 2.0 Array 1460479_PM_at - -
          Mouse Genome 430 2.0 Array 1460479_at - -
          Mouse Expression Set 430 1460479_at - -
          Mouse Expression Set 430 1460479_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460479_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460479_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 356777 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104590746 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0